2-Methylimidazo[1,2-a]pyridin 97%
Lieferant: Apollo Scientific
OR110181-1G
APOSOR110181-1GEA
79.9
EUR
APOSOR110181-1G
2-Methylimidazo[1,2-a]pyridin 97%
2-Methylimidazo[1,2-a]pyridin
Formel:
C₈H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02251330 CAS-Nummer: 934-37-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...