6-Bromo-2-methylimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific
Gefahr

OR110182-1G
APOSOR110182-1GEA 85.7 EUR
APOSOR110182-1G
6-Bromo-2-methylimidazo[1,2-a]pyridine
6-Bromo-2-methylimidazo[1,2-a]pyridine
Formel: C₈H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09800594
CAS-Nummer: 4044-99-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR