2-Amino-7-bromchinazolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromoquinazolin-2-amine

OR110183-1G
APOSOR110183-1GEA 109 EUR
APOSOR110183-1G
2-Amino-7-bromchinazolin
2-Amino-7-bromoquinazoline
Formel: C₈H₆BrN₃
Molecular Weight: 224,0600 g/mol
Schmelzpunkt: 280…283 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027698
CAS-Nummer: 190274-15-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR