4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one

Lieferant: Apollo Scientific

Synonyme: 4-(3-Nitro-4-piperidin-1-ylphenyl)phthalazin-1(2H)-one

OR110185-1G
APOSOR110185-1GEA 0 EUR
APOSOR110185-1G
4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one
4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one
Formel: C₁₉H₁₈N₄O₃
Molecular Weight: 350,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00116203
CAS-Nummer: 218144-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR