Dimethyl-2-(4-nitrophenyl)malonate

Lieferant: Apollo Scientific

Synonyme: Dimethyl 2-(4-nitrophenyl)malonate

OR110186-1G
APOSOR110186-1GEA 0 EUR
APOSOR110186-1G
Dimethyl-2-(4-nitrophenyl)malonate
Dimethyl-2-(4-nitrophenyl)malonate
Formel: C₁₁H₁₁NO₆
Molecular Weight: 253,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425806
CAS-Nummer: 4033-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR