3-Chlorimidazo[1,2-a]pyridin-6-carbonsäure

Lieferant: Apollo Scientific
Achtung

OR110190-1G
APOSOR110190-1GEA 101 EUR
APOSOR110190-1G
3-Chlorimidazo[1,2-a]pyridin-6-carbonsäure
3-Chlorimidazo[1,2-a]pyridin-6-carbonsäure
Formel: C₈H₅ClN₂O₂
Molecular Weight: 196.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07021510
CAS-Nummer: 900019-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR