3-Chloroimidazo[1,2-a]pyridine-6-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR110191-1G
APOSOR110191-1GEA 219 EUR
APOSOR110191-1G
3-Chloroimidazo[1,2-a]pyridine-6-carbonitrile
3-Chloroimidazo[1,2-a]pyridine-6-carbonitrile
Formel: C₈H₄ClN₃
Molecular Weight: 177,59 g/mol
Schmelzpunkt: 199…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09994775
CAS-Nummer: 1019027-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR