N,N-Dimethyl-N-[2-(3-nitropyridin-4-yl)vinyl]amine

Lieferant: Apollo Scientific

OR110201-1G
APOSOR110201-1GEA 0 EUR
APOSOR110201-1G
N,N-Dimethyl-N-[2-(3-nitropyridin-4-yl)vinyl]amine
N,N-Dimethyl-N-[2-(3-nitropyridin-4-yl)vinyl]amine
Formel: C₉H₁₁N₃O₂
Molecular Weight: 193,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505062
CAS-Nummer: 64679-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR