3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one

Lieferant: Apollo Scientific

OR110202-1G
APOSOR110202-1GEA 128 EUR
APOSOR110202-1G
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one
Formel: C₉H₁₂N₂OS
Molecular Weight: 196,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056641
CAS-Nummer: 886361-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR