2-(5-Methyl-1H-pyrazol-3-yl)-1,3-thiazole

Lieferant: Apollo Scientific
Achtung

OR110203-1G
APOSOR110203-1GEA 434 EUR
APOSOR110203-1G
2-(5-Methyl-1H-pyrazol-3-yl)-1,3-thiazole
2-(5-Methyl-1H-pyrazol-3-yl)-1,3-thiazole
Formel: C₇H₇N₃S
Molecular Weight: 165.22 g/mol
Schmelzpunkt: 116…118 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056642
CAS-Nummer: 1187559-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR