3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one

Lieferant: Apollo Scientific

Synonyme: 3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-propen-1-one

OR110204-1G
APOSOR110204-1GEA 81.2 EUR
APOSOR110204-1G
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one
3-(Dimethylamino)-1-(1,3-thiazol-2-yl)prop-2-en-1-one
Formel: C₈H₁₀N₂OS
Molecular Weight: 182,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00105071
CAS-Nummer: 191729-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR