Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-amino-4-chlorobenzo[b]thiophene-2-carboxylate

OR110209-1G
APOSOR110209-1GEA 49.3 EUR
APOSOR110209-1G
Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate
Ethyl 3-amino-4-chloro-1-benzothiophene-2-carboxylate
Formel: C₁₁H₁₀ClNO₂S
Molecular Weight: 255.721 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00661179
CAS-Nummer: 67189-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR