Ethyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110211-1G
APOSOR110211-1GEA 395 EUR
APOSOR110211-1G
Ethyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate
Ethyl 3-amino-4-bromobenzo[b]thiophene-2-carboxylate
Formel: C₁₁H₁₀BrNO₂S
Molecular Weight: 300,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615812
CAS-Nummer: 1227954-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR