3-Iodo-1H-indole-2-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR110212-250MG
APOSOR110212-250MGEA
21.9
EUR
APOSOR110212-250MG
3-Iodo-1H-indole-2-carbohydrazide
3-Iodo-1H-indole-2-carbohydrazide
Formel:
C₉H₈IN₃O Molecular Weight: 301,084 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11615813 CAS-Nummer: 1227954-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...