3-Iodo-1H-indole-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110213-1G
APOSOR110213-1GEA 232 EUR
APOSOR110213-1G
3-Iodo-1H-indole-2-carboxylic acid
3-Iodo-1H-indole-2-carboxylic acid
Formel: C₉H₆INO₂
Molecular Weight: 287,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657947
CAS-Nummer: 167631-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR