Methyl 2-ethyl-1,3-benzoxazole-7-carboxylate

Lieferant: Apollo Scientific

OR110218-1G
APOSOR110218-1GEA 232 EUR
APOSOR110218-1G
Methyl 2-ethyl-1,3-benzoxazole-7-carboxylate
Methyl 2-ethyl-1,3-benzoxazole-7-carboxylate
Formel: C₁₁H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11615816
CAS-Nummer: 1227955-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR