3-(Dimethylamino)-1-(5-methyl-3-phenylisoxazol-4-yl)prop-2-en-1-one

Lieferant: Apollo Scientific

OR110219-1G
APOSOR110219-1GEA 92.8 EUR
APOSOR110219-1G
3-(Dimethylamino)-1-(5-methyl-3-phenylisoxazol-4-yl)prop-2-en-1-one
3-(Dimethylamino)-1-(5-methyl-3-phenylisoxazol-4-yl)prop-2-en-1-one
Formel: C₁₅H₁₆N₂O₂
Molecular Weight: 256,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662706
CAS-Nummer: 265125-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR