7-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110224-1G
APOSOR110224-1GEA 202 EUR
APOSOR110224-1G
7-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
7-Chloroimidazo[1,2-a]pyridine-2-carbohydrazide
Formel: C₈H₇ClN₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD11505066
CAS-Nummer: 1215727-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR