5-Piperidin-1-yl-1,3,4-thiadiazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR110229-1G
APOSOR110229-1GEA 174 EUR
APOSOR110229-1G
5-Piperidin-1-yl-1,3,4-thiadiazol-2-amine
5-Piperidin-1-yl-1,3,4-thiadiazol-2-amine
Formel: C₇H₁₂N₄S
Molecular Weight: 184,262 g/mol
Schmelzpunkt: 205,5…206.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08273069
CAS-Nummer: 71125-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR