5-Pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR110230-1G
APOSOR110230-1GEA 203 EUR
APOSOR110230-1G
5-Pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine
5-Pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine
Formel: C₆H₁₀N₄S
Molecular Weight: 170,235 g/mol
Schmelzpunkt: 226,5…228.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06243032
CAS-Nummer: 71125-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR