6-Iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR110232-1G
APOSOR110232-1GEA 235 EUR
APOSOR110232-1G
6-Iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid
6-Iodo-8-methylimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₉H₇IN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11846256
CAS-Nummer: 1227954-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR