2-Butyl-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
OR110234-1G
APOSOR110234-1GEA
69.6
EUR
APOSOR110234-1G
2-Butyl-1H-benzimidazole
2-Butyl-1H-benzimidazole
Formel:
C₁₁H₁₄N₂ Molecular Weight: 174,25 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00714456 CAS-Nummer: 5851-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...