2-Propyl-1H-benzo[d]imidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Propylbenzimidazole

OR110235-5G OR110235-10G
APOSOR110235-5GEA 121 EUR
APOSOR110235-5G APOSOR110235-10G
2-Propyl-1H-benzo[d]imidazole
2-Propyl-1H-benzo[d]imidazole
Formel: C₁₀H₁₂N₂
Molecular Weight: 160.2200 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00087477
CAS-Nummer: 5465-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR