(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one

Lieferant: Apollo Scientific

OR110236-25G
APOSOR110236-25GEA 69.6 EUR
APOSOR110236-25G
(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one
(2E)-1-(4-Bromophenyl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₁₁H₁₂BrNO
Molecular Weight: 254,13 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00121187
CAS-Nummer: 73387-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR