4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Lieferant: Apollo Scientific
OR110244-1G
OR110244-250MG
APOSOR110244-1GEA
221
EUR
APOSOR110244-1G
APOSOR110244-250MG
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Formel:
C₉H₈N₂O₄ Molecular Weight: 208.171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12406815 CAS-Nummer: 132522-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...