2-Methyl-1,3-benzoxazole-5-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylbenzo[d]oxazole-5-carboxylic acid

OR110248-5G
APOSOR110248-5GEA 85.7 EUR
APOSOR110248-5G
2-Methyl-1,3-benzoxazole-5-carboxylic acid 95%
2-Methyl-1,3-benzoxazole-5-carboxylic acid
Formel: C₉H₇NO₃
Molecular Weight: 177.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09759125
CAS-Nummer: 90322-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR