1-(4-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR110250-1G
APOSOR110250-1GEA 116 EUR
APOSOR110250-1G
1-(4-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Bromophenyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₀BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00448856
CAS-Nummer: 91348-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR