Methyl 2-chloro-6-(4-methylpiperazin-1-yl)isonicotinate

Lieferant: Apollo Scientific

OR110255-1G
APOSOR110255-1GEA 206 EUR
APOSOR110255-1G
Methyl 2-chloro-6-(4-methylpiperazin-1-yl)isonicotinate
Methyl 2-chloro-6-(4-methylpiperazin-1-yl)isonicotinate
Formel: C₁₂H₁₆ClN₃O₂
Molecular Weight: 269,727 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756247
CAS-Nummer: 1227954-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR