Quinoxaline-6-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110256-1G
APOSOR110256-1GEA 130 EUR
APOSOR110256-1G
Quinoxaline-6-carbohydrazide
Quinoxaline-6-carbohydrazide
Formel: C₉H₈N₄O
Molecular Weight: 188.186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114841
CAS-Nummer: 175204-23-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR