2-Amino-6-bromchinazolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromoquinazolin-2-amine

OR110259-1G
APOSOR110259-1GEA 78.3 EUR
APOSOR110259-1G
2-Amino-6-bromchinazolin
2-Amino-6-bromoquinazoline
Formel: C₈H₆BrN₃
Molecular Weight: 224,0600 g/mol
Schmelzpunkt: 270…272 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04114089
CAS-Nummer: 190273-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR