Ethyl 1-methyl-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific

OR110264-1G OR110264-5G
APOSOR110264-1GEA 92.8 EUR
APOSOR110264-1G APOSOR110264-5G
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Formel: C₈H₁₁NO₂
Molecular Weight: 153.178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179573
CAS-Nummer: 23466-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR