1-Methyl-1H-pyrrole-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110266-1G
APOSOR110266-1GEA 139 EUR
APOSOR110266-1G
1-Methyl-1H-pyrrole-2-carbohydrazide
1-Methyl-1H-pyrrole-2-carbohydrazide
Formel: C₆H₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00141854
CAS-Nummer: 113398-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR