Methyl-2-aminonicotinat

Lieferant: Apollo Scientific

Synonyme: Methyl 2-aminonicotinate

OR110267-100G
APOSOR110267-100GEA 143 EUR
APOSOR110267-100G
Methyl-2-aminonicotinat
Methyl-2-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00128872
CAS-Nummer: 14667-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR