4-Methylphenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane

OR11027-5G
APOSOR11027-5GEA 21.9 EUR
APOSOR11027-5G
4-Methylphenylboronsäurepinakolester
4-Methylphenylboronsäurepinakolester
Formel: C₁₃H₁₉BO₂
Molecular Weight: 218,1 g/mol
Schmelzpunkt: 54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453662
CAS-Nummer: 195062-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR