Methyl 4-amino-3-bromo-5-methylbenzoate

Lieferant: Apollo Scientific
Achtung

OR110270-10G OR110270-1G OR110270-5G
APOSOR110270-10GEA 89.9 EUR
APOSOR110270-10G APOSOR110270-1G APOSOR110270-5G
Methyl 4-amino-3-bromo-5-methylbenzoate
Methyl 4-amino-3-bromo-5-methylbenzoate
Formel: C₉H₁₀BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07021337
CAS-Nummer: 900019-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR