Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110272-1G OR110272-5G
APOSOR110272-1GEA 232 EUR
APOSOR110272-1G APOSOR110272-5G
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Formel: C₁₁H₁₁NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD08706234
CAS-Nummer: 35212-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR