4-Bromo-1-methyl-1H-indazol-3-amine

Lieferant: Apollo Scientific

OR110273-1G
APOSOR110273-1GEA 364 EUR
APOSOR110273-1G
4-Bromo-1-methyl-1H-indazol-3-amine
4-Bromo-1-methyl-1H-indazol-3-amine
Formel: C₈H₈BrN₃
Molecular Weight: 226,073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847110
CAS-Nummer: 1260854-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR