Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110275-1G
APOSOR110275-1GEA 232 EUR
APOSOR110275-1G
Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Ethyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Formel: C₁₂H₁₃NO₃S
Molecular Weight: 251,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194590
CAS-Nummer: 77373-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR