(3-Bromoimidazo[1,2-a]pyridin-6-yl)methanol

Lieferant: Apollo Scientific
Achtung

OR110280-1G
APOSOR110280-1GEA 330 EUR
APOSOR110280-1G
(3-Bromoimidazo[1,2-a]pyridin-6-yl)methanol
(3-Bromoimidazo[1,2-a]pyridin-6-yl)methanol
Formel: C₈H₇BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13191321
CAS-Nummer: 1004550-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR