Methyl 2-ethoxy-5-iodonicotinate

Lieferant: Apollo Scientific
Achtung

OR110282-1G
APOSOR110282-1GEA 346 EUR
APOSOR110282-1G
Methyl 2-ethoxy-5-iodonicotinate
Methyl 2-ethoxy-5-iodonicotinate
Formel: C₉H₁₀INO₃
Molecular Weight: 307,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194591
CAS-Nummer: 335078-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR