2-Acetyl-6-iodo-2H-indazole

Lieferant: Apollo Scientific
Achtung

OR110286-1G
APOSOR110286-1GEA 319 EUR
APOSOR110286-1G
2-Acetyl-6-iodo-2H-indazole
2-Acetyl-6-iodo-2H-indazole
Formel: C₉H₇IN₂O
Molecular Weight: 286,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14583146
CAS-Nummer: 1305711-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR