Methyl 2-acetyl-2H-indazole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR110287-1G
APOSOR110287-1GEA
267
EUR
APOSOR110287-1G
Methyl 2-acetyl-2H-indazole-4-carboxylate
Methyl 2-acetyl-2H-indazole-4-carboxylate
Formel:
C₁₁H₁₀N₂O₃ Molecular Weight: 218,209 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14635899 CAS-Nummer: 1303890-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...