4-Ethyl-N-methylbenzenesulphonamide
Lieferant: Apollo Scientific
OR110290-1G
OR110290-250MG
OR110290-5G
APOSOR110290-1GEA
0
EUR
APOSOR110290-1G
APOSOR110290-250MG
APOSOR110290-5G
4-Ethyl-N-methylbenzenesulphonamide
4-Ethyl-N-methylbenzenesulphonamide
Formel:
C₉H₁₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD03550606 CAS-Nummer: 401583-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...