4-Ethyl-N-methylbenzenesulphonamide

Lieferant: Apollo Scientific

OR110290-1G OR110290-250MG OR110290-5G
APOSOR110290-1GEA 0 EUR
APOSOR110290-1G APOSOR110290-250MG APOSOR110290-5G
4-Ethyl-N-methylbenzenesulphonamide
4-Ethyl-N-methylbenzenesulphonamide
Formel: C₉H₁₃NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03550606
CAS-Nummer: 401583-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR