Methyl 1,3-benzoxazole-6-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl benzo[d]oxazole-6-carboxylate

OR110295-1G
APOSOR110295-1GEA 127 EUR
APOSOR110295-1G
Methyl 1,3-benzoxazole-6-carboxylate 95%
Methyl 1,3-benzoxazole-6-carboxylate
Formel: C₉H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14635902
CAS-Nummer: 1305711-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR