4-Chloro-N-ethylbenzenesulphonamide

Lieferant: Apollo Scientific

OR110301-1G
APOSOR110301-1GEA 0 EUR
APOSOR110301-1G
4-Chloro-N-ethylbenzenesulphonamide
4-Chloro-N-ethylbenzenesulphonamide
Formel: C₈H₁₀ClNO₂S
Molecular Weight: 219.689 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01211969
CAS-Nummer: 6318-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR