6-Bromobenzo[d]isoxazol-3-amine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR110302-1G
APOSOR110302-1GEA
72.5
EUR
APOSOR110302-1G
6-Bromobenzo[d]isoxazol-3-amine 97%
6-Bromobenzo[d]isoxazol-3-amine
Formel:
C₇H₅BrN₂O Schmelzpunkt: 114…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09788587 CAS-Nummer: 177995-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...