Methyl 2-amino-5-chloronicotinate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-amino-5-chloropyridine-3-carboxylate, 2-Amino-5-chloro-3-(methoxycarbonyl)pyridine
OR110303-5G
APOSOR110303-5GEA
120
EUR
APOSOR110303-5G
Methyl 2-amino-5-chloronicotinate 95%
Methyl 2-amino-5-chloronicotinate
Formel:
C₇H₇ClN₂O₂ Molecular Weight: 186,596 g/mol Schmelzpunkt: 133…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09031678 CAS-Nummer: 50735-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...