1H-Indazole-4-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110304-1G
APOSOR110304-1GEA 231 EUR
APOSOR110304-1G
1H-Indazole-4-carbohydrazide
1H-Indazole-4-carbohydrazide
Formel: C₈H₈N₄O
Molecular Weight: 176,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506090
CAS-Nummer: 1086392-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR