Ethyl 3-amino-5-bromo-1-benzothiophene-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR110309-1G
APOSOR110309-1GEA
58
EUR
APOSOR110309-1G
Ethyl 3-amino-5-bromo-1-benzothiophene-2-carboxylate
Ethyl 3-amino-5-bromo-1-benzothiophene-2-carboxylate
Formel:
C₁₁H₁₀BrNO₂S Molecular Weight: 300,172 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14635904 CAS-Nummer: 1308649-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...